4-Ethynylaniline 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(4-Aminophenyl)acetylene, 1-Ethynyl-4-aminobenzene, 1-Amino-4-ethynylbenzene, (4-Aminophenyl)ethyne
432950010
432950050
ACRO432950010EA
83.8
EUR
ACRO432950010
ACRO432950050
4-Ethynylaniline 97%
4-Ethynylaniline
Formula:
C₈H₇N MW: 117.15 g/mol Melting Pt: 100…102 °C Storage Temperature: Freezer |
MDL Number:
MFCD00168824 CAS Number: 14235-81-5 |
Specification Test Results
Appearance | Yellow to brown Powder |
Infrared spectrum | Conforms |
Melting point | 98 to 104 °C |
GC | ≥96.0 % |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.4 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...