2-(2-Chlorophenyl)glycine 98%

Supplier: Thermo Fisher Scientific
Warning

433340250 433341000
ACRO433340250EA 115 EUR
ACRO433340250 ACRO433341000
2-(2-Chlorophenyl)glycine 98%
2-(2-Chlorophenyl)glycine
Formula: C₈H₈ClNO₂
Melting Pt: 182,9…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00049324
CAS Number: 88744-36-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR