2,3-Dibromopropionyl chloride 97%

Supplier: Thermo Scientific
Danger

Synonyms: 2,3-Dibromopropionylchloride

433410050 433410250
ACRO433410050EA 120 EUR
ACRO433410050 ACRO433410250
2,3-Dibromopropionyl chloride 97%
2,3-Dibromopropionyl chloride
Formula: C₃H₃Br₂ClO
MW: 250,31 g/mol
Density: 2,14 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000712
CAS Number: 18791-02-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR