Cysteamine 95%

Supplier: Thermo Fisher Scientific
Danger

434710050 434710250
ACRO434710050EA 98.5 EUR
ACRO434710050 ACRO434710250
Cysteamine 95%
Cysteamine
Formula: C₂H₇NS
Melting Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008196
CAS Number: 60-23-1
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 94 to 99 °C
Titration Iodimetric ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR