5-Iodo-1H-indazole 95%

Supplier: Thermo Fisher Scientific
Warning

435710010 435710050
ACRO435710010EA 164 EUR
ACRO435710010 ACRO435710050
5-Iodo-1H-indazole 95%
5-Iodo-1H-indazole
Formula: C₇H₅IN₂
MW: 244.0300 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD07781642
CAS Number: 55919-82-9

Order Now

Specification Test Results

Appearance Yellow to brown to purple Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 156 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR