2-Bromo-5-hydroxypyrazine 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Bromopyrazin-2-ol
435890010
435890050
ACRO435890010EA
298
EUR
ACRO435890010
ACRO435890050
2-Bromo-5-hydroxypyrazine 97%
2-Bromo-5-hydroxypyrazine
Formula:
C₄H₃BrN₂O MW: 174,9800 g/mol Melting Pt: 115…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD06245330 CAS Number: 374063-92-0 |
Specification Test Results
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
GC | ≥98.5 % |
Specific optical rotation | +67° to +69° (20°C, 589 nm) (c=2.3, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...