3-Chloro-4-methylbenzaldehyde 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
436690010
436690050
ACRO436690010EA
58.5
EUR
ACRO436690010
ACRO436690050
3-Chloro-4-methylbenzaldehyde 97%
3-Chloro-4-methylbenzaldehyde
Formula:
C₈H₇ClO Boiling Pt: 232 °C (760 mmHg) Melting Pt: 37…31 °C Storage Temperature: Ambient |
MDL Number:
MFCD06411284 CAS Number: 3411-03-8 |
Specification Test Results
Appearance | Colorless to white Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...