N,N-Diethylsalicylamide 97%

Supplier: Thermo Fisher Scientific
Warning

437250050 437250250
ACRO437250050EA 110 EUR
ACRO437250050 ACRO437250250
N,N-Diethylsalicylamide 97%
N,N-Diethylsalicylamide
Formula: C₁₁H₁₅NO₂
Melting Pt: 99,1…100.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00010816
CAS Number: 19311-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR