5-Phenyl-2-furaldehyde 96%
Supplier: Thermo Scientific
|
Warning
|
437310010
437310050
ACRO437310010EA
71.9
EUR
ACRO437310010
ACRO437310050
5-Phenyl-2-furaldehyde 96%
5-Phenyl-2-furaldehyde
Formula:
C₁₁H₈O₂ Boiling Pt: 104…108 °C (6 mmHg) Melting Pt: 29…33 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01910161 CAS Number: 13803-39-9 |
Specification Test Results
Appearance | Yellow to green to brown Solid or liquid |
Infrared spectrum | Conforms |
GC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...