4-Amino-3-formylpyridine 95%

Supplier: Thermo Fisher Scientific
Warning

437350010 437350050
ACRO437350010EA 150 EUR
ACRO437350010 ACRO437350050
4-Amino-3-formylpyridine 95%
4-Amino-3-formylpyridine
Formula: C₆H₆N₂O
Melting Pt: 111,2…112.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038972
CAS Number: 42373-30-8

Order Now

Specification Test Results

Appearance Yellow to brown Powder or solid
Infrared spectrum Conforms
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR