(2-Chloropyridin-4-yl)methylamine 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (2-Chloropyridin-4-yl)methanamine, 4-Aminomethyl-2-chloropyridine

438890010 438890050
ACRO438890010EA 222 EUR
ACRO438890010 ACRO438890050
(2-Chloropyridin-4-yl)methylamine 95%
(2-Chloropyridin-4-yl)methylamine
Formula: C₆H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD06212875
CAS Number: 144900-57-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR