5-Bromo-2-methoxypyrimidine 97%
Supplier: Thermo Scientific
|
Warning
|
441850010
441850050
ACRO441850010EA
101
EUR
ACRO441850010
ACRO441850050
5-Bromo-2-methoxypyrimidine 97%
5-Bromo-2-methoxypyrimidine
Formula:
C₅H₅BrN₂O MW: 189,01 g/mol Melting Pt: 57…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973351 CAS Number: 14001-66-2 |
Specification Test Results
Appearance | White to light brown Crystalline powder or solid |
Infrared spectrum | Conforms |
Melting point | 55 to 59 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...