Picolinamide 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Pyridinecarboxamide, Pyridine-2-carboxamide
443360100
443360500
ACRO443360100EA
113
EUR
ACRO443360100
ACRO443360500
Picolinamide 98%
Picolinamide
Formula:
C₆H₆N₂O MW: 122,13 g/mol Boiling Pt: 143 °C (2,7kPa) Melting Pt: 106…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023483 CAS Number: 1452-77-3 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...