4-Iodo-1H-pyrazol-5-amine 97%

Supplier: Thermo Scientific
Warning

444520010 444520050
ACRO444520010EA 129 EUR
ACRO444520010 ACRO444520050
4-Iodo-1H-pyrazol-5-amine 97%
4-Iodo-1H-pyrazol-5-amine
Formula: C₃H₄IN₃
Melting Pt: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD22422021
CAS Number: 81542-51-0

Order Now

Specification Test Results

Appearance Off-white to brown Solid
Infrared spectrum Conforms
Melting point 140 to 146 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR