3-Iodothiophene 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
443380010
443380050
ACRO443380010EA
71.9
EUR
ACRO443380010
ACRO443380050
3-Iodothiophene 97%
3-Iodothiophene
Formula:
C₄H₃IS Boiling Pt: 195 °C (760 mmHg) Melting Pt: –13 °C Density: 2,066 g/cm³ Flash Pt: 78 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037901 CAS Number: 10486-61-0 |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...