6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Carbamic acid,N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinyl]-,1,1-dimethylethyl ester, tert-Butyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate, 2-(BOC-Amino)pyridine-5-boronic acid, pinacol ester
444660010
444660050
ACRO444660010EA
148
EUR
ACRO444660010
ACRO444660050
6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester 97%
6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester
Formula:
C₁₆H₂₅BN₂O₄ MW: 320,196 g/mol Melting Pt: 203…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD07781162 CAS Number: 910462-31-6 |
Specification Test Results
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Melting point | 202 to 208 °C |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...