Trichloroacetic anhydride 95%

Supplier: Thermo Scientific
Danger

446090250 446091000
ACRO446090250EA 98.9 EUR
ACRO446090250 ACRO446091000
Trichloroacetic anhydride 95%
Trichloroacetic anhydride
Formula: C₄Cl₆O₃
MW: 308,76 g/mol
Boiling Pt: 139…141 °C (60 mmHg)
Melting Pt: –2 °C
Density: 1,69 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000793
CAS Number: 4124-31-6
UN: 3265
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4870 (20°C, 589 nm)
Titration by coupling ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR