Pyridazine-3-carboxylic acid 97%
Supplier: Thermo Scientific
|
Warning
|
446720010
446720050
ACRO446720010EA
101
EUR
ACRO446720010
ACRO446720050
Pyridazine-3-carboxylic acid 97%
Pyridazine-3-carboxylic acid
Formula:
C₅H₄N₂O₂ MW: 124,10 g/mol Melting Pt: 190,8…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646333 CAS Number: 2164-61-6 |
Specification Test Results
Appearance | Off-white to yellow to light brown Powder or solid |
Infrared spectrum | Conforms |
Water | ≤1.5 % |
NMR (1H-NMR) | Consistent with structure |
Titration with NaOH | ≥96.0 to ≤104.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...