Ethyl azidoacetate 25% in toluene

Supplier: Thermo Scientific
Danger

Synonyms: (2-ethoxy-2-oxoethyl)imino-iminoammonium, Ethyl 2-(1H-triazirin-1-yl)acetate, Acetic acid, 2-azido-, ethyl ester

448780050 448780250
ACRO448780050EA 52.8 EUR
ACRO448780050 ACRO448780250
Ethyl azidoacetate 25% in toluene
Ethyl azidoacetate
Formula: C₄H₇N₃O₂
MW: 129,12 g/mol
Density: 0,92 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00190177
CAS Number: 637-81-0
UN: 1294
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
NMR ≥24 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR