Diisopropyl methylphosphonate 90%

Supplier: Thermo Scientific
Warning

449380050 449380250 449381000
ACRO449380050EA 44.4 EUR
ACRO449380050 ACRO449380250 ACRO449381000
Diisopropyl methylphosphonate 90%
Diisopropyl methylphosphonate
Formula: C₇H₁₇O₃P
Boiling Pt: 50…51 °C (1 mmHg)
Density: 0,985 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00015022
CAS Number: 1445-75-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4130 (20°C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR