(-)-Epigallocatechin gallate 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG
449010100
449010500
ACRO449010100EA
67.7
EUR
ACRO449010100
ACRO449010500
(-)-Epigallocatechin gallate 95%
(-)-Epigallocatechin gallate
Formula:
C₂₂H₁₈O₁₁ MW: 458.38 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00075940 CAS Number: 989-51-5 |
Specification Test Results
Appearance | White to light pink Powder |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...