3-Amino-1-bromoisoquinoline 97%
Supplier: Thermo Scientific
|
Warning
|
449160010
449160050
ACRO449160010EA
223
EUR
ACRO449160010
ACRO449160050
3-Amino-1-bromoisoquinoline 97%
3-Amino-1-bromoisoquinoline
Formula:
C₉H₇BrN₂ Melting Pt: 144…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00102193 CAS Number: 13130-79-5 |
Specification Test Results
Appearance | Yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 143 to 147 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...