N-Boc-4-oxo-L-proline 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Boc-4-oxo-Pro-OH, (S)-1-Boc-4-oxo-2-pyrrolidinecarboxylic Acid, (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, (S)-1-(tert-Butoxycarbonyl)-4-oxo-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4-oxo-L-proline
449280010
449280050
ACRO449280010EA
140
EUR
ACRO449280010
ACRO449280050
N-Boc-4-oxo-L-proline 98%
N-Boc-4-oxo-L-proline
Formula:
C₁₀H₁₅NO₅ MW: 229,23 g/mol Melting Pt: 155…163 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01860669 CAS Number: 84348-37-8 |
Specification Test Results
Appearance | White to tan Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 157 to 163 °C |
HPLC | ≥97.5 % |
Specific optical rotation | +18° to +23° (20 °C, 589 nm) (c=0.5, acetone) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...