(S)-(-)-Epoxystyrene 94%

Supplier: Thermo Fisher Scientific
Danger

449450050 449450250
ACRO449450050EA 157 EUR
ACRO449450050 ACRO449450250
(S)-(-)-Epoxystyrene 94%
(S)-(-)-Epoxystyrene
Formula: C₈H₈O
Boiling Pt: 192…194 °C
Melting Pt: –37 °C
Density: 1,051 g/cm³
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064310
CAS Number: 20780-54-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5360 (20°C, 589 nm)
Water ≤0.5 %
GC ≥92 %
Specific optical rotation -31° to -35° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR