5-Hexenoic acid 98%

Supplier: Thermo Scientific
Danger

449520010 449520050
ACRO449520010EA 68 EUR
ACRO449520010 ACRO449520050
5-Hexenoic acid 98%
5-Hexenoic acid
Formula: C₆H₁₀O₂
Boiling Pt: 203 °C (760 mmHg)
Melting Pt: -37 °C
Density: 0,961 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00046558
CAS Number: 1577-22-6
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR