2-Quinolinecarbonitrile 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Quinoline-2-carbonitrile
449580010
449580050
ACRO449580010EA
92.2
EUR
ACRO449580010
ACRO449580050
2-Quinolinecarbonitrile 97%
2-Quinolinecarbonitrile
Formula:
C₁₀H₆N₂ MW: 154,17 g/mol Boiling Pt: 324 °C (1013 hPa) Melting Pt: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134341 CAS Number: 1436-43-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to yellow to beige Powder |
Infrared spectrum | Conforms |
Melting point | 92 to 96 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...