3-Methylstyrene 95%

Supplier: Thermo Scientific
Danger

Synonyms: 3-Vinyltoluene

450870010 450870050
ACRO450870010EA 33.3 EUR
ACRO450870010 ACRO450870050
3-Methylstyrene 95%
3-Methylstyrene
Formula: C₉H₁₀
MW: 118,18 g/mol
Boiling Pt: 170…171 °C
Melting Pt: 82…81 °C
Density: 0,902 g/cm³ (18 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00008617
CAS Number: 100-80-1
UN: 2618
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Stabilizer 0.1 % 4-tert-Butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR