3,4-Dimethylthiophene 97%
Supplier: Thermo Scientific
|
Warning
|
450950010
450950050
ACRO450950010EA
153
EUR
ACRO450950010
ACRO450950050
3,4-Dimethylthiophene 97%
3,4-Dimethylthiophene
Formula:
C₆H₈S MW: 112,1900 g/mol Boiling Pt: 144 °C (760 mmHg) Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD01764731 CAS Number: 632-15-5 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to yellow (may darken on storage) Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...