3-Oxetanamine 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-amino-oxetane, 3-Aminooxetane
451212500
451215000
ACRO451212500EA
147
EUR
ACRO451212500
ACRO451215000
3-Oxetanamine 95%
3-Oxetanamine
Formula:
C₃H₇NO MW: 73,09 g/mol Boiling Pt: 99 °C Flash Pt: 24 °C Storage Temperature: Freezer |
MDL Number:
MFCD08544363 CAS Number: 21635-88-1 UN: 1992 ADR: 3,III |
Specification Test Results
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...