5-Amino-1-methyl-1H-pyrazole 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole
452610250
452611000
ACRO452610250EA
253
EUR
ACRO452610250
ACRO452611000
5-Amino-1-methyl-1H-pyrazole 97%
5-Amino-1-methyl-1H-pyrazole
Formula:
C₄H₇N₃ MW: 97,12 g/mol Melting Pt: 73…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068156 CAS Number: 1192-21-8 |
Specification Test Results
Appearance | White to yellow Solid |
Infrared spectrum | Conforms |
Melting point | 69 to 77 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...