2,4-Diacetylphloroglucinol 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1,1'-(2,4,6-Trihydroxy-1,3-phenylene)bisethanone, 1-(3-acetyl-2,4,6-trihydroxyphenyl)ethan-1-one, Ethanone,1,1'-(2,4,6-trihydroxy-1,3-phenylene)bis-
452800010
452800050
ACRO452800010EA
255
EUR
ACRO452800010
ACRO452800050
2,4-Diacetylphloroglucinol 97%
2,4-Diacetylphloroglucinol
Formula:
C₁₀H₁₀O₅ MW: 210.19 g/mol Melting Pt: 164…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100483 CAS Number: 2161-86-6 |
Specification Test Results
Appearance | Off-white to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...