4-(2-Pyridylazo)resorcinol 97+% ACS
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-(2'-Pyridylazo)resorcinol
452930010
452930050
ACRO452930010EA
28.9
EUR
ACRO452930010
ACRO452930050
4-(2-Pyridylazo)resorcinol 97+% ACS
4-(2-Pyridylazo)resorcinol
Formula:
C₁₁H₉N₃O₂ MW: 215,21 g/mol Melting Pt: 188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006256 CAS Number: 1141-59-9 |
Specification Test Results
Appearance | Orange Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.0 % |
Suitability | (as complexom. indicator) Passes test |
Clarity of solution | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...