2,3,5-Trimethylfuran 99%

Supplier: Thermo Scientific
Danger

Synonyms: Furan, 2,3,5-trimethyl-

454470010 454470050
ACRO454470010EA 124 EUR
ACRO454470010 ACRO454470050
2,3,5-Trimethylfuran 99%
2,3,5-Trimethylfuran
Formula: C₇H₁₀O
MW: 110,16 g/mol
Boiling Pt: 121…122 °C
Density: 0,89 g/cm³
Flash Pt: 20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03931179
CAS Number: 10504-04-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR