3'-Azido-3'-deoxythymidine 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymine, Zidovudine, Azidothymidine, AZT
454980010
454980050
454980250
ACRO454980010EA
49
EUR
ACRO454980010
ACRO454980050
ACRO454980250
3'-Azido-3'-deoxythymidine 98%
3'-Azido-3'-deoxythymidine
Formula:
C₁₀H₁₃N₅O₄ MW: 267,25 g/mol Melting Pt: 118…120 °C Storage Temperature: Freezer |
MDL Number:
MFCD00006536 CAS Number: 30516-87-1 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 122 to 126 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...