Emulating indicators, TST Control™
Supplier: BROWNE HEALTH CARE
3726 2342
BROH3726EA 112 EUR
BROH3726 BROH2342
Emulating indicators, TST Control™
Sterilisation Indicators
When placed inside trays, packs or pouches, a TST Control™ emulating indicator will confirm that good quality steam has penetrated to the point of placement.