Cut resistant gloves, KryTech 557
Supplier: MAPA
34557041 34557047 34557048 34557049 34557040 34557046
MAPA34557041EA 17 EUR
MAPA34557041 MAPA34557047 MAPA34557048 MAPA34557049 MAPA34557040 MAPA34557046
Cut resistant gloves, KryTech 557
Gloves
These gloves made of HDPE fibres with PU coating on palm and fingertips provide excellent abrasion resistance and good cut protection. Suitable for intricate handlings in reasonably clean environments.
  • Seamless knitting
  • Suitable for long-term wear
  • Excellent dexterity: anatomical shape
  • Ergonomic to reduce hand fatigue
  • Reinforced welt in nitrile for an improved durability for the most exposed areas
Typical applications include: Handling and sorting small, sharp parts, handling cardboard and paper, handling metal parts, laying tiles, inserts, frames or zinc sheeting, installing steel structures and screwing.
Certifications: EN 388:2016 (4343B); ISO 13997: 5.3N (540 g); Cat. II