β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Supplier: MERCK PRODUCTION CHEMICALS
1.37074.0250
1.37074.1000
1.37074.0250EA
498
EUR
1.37074.0250
1.37074.1000
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
2-Morpholinoethanesulfonic acid monohydrate (MES) is suitable for biopharmaceutical production EMPROVE® bio.
Formula:
C₆H₁₃NO₄S·1H₂O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149409 CAS Number: 145224-94-8 EINECS: 224-632-3 |
Specification Test Results
Synonyms | MES |
pH value | 3,6 (10 g/l, H2O, 20 °C) |
Bulk density | 570 kg/m3 |
Solubility | 80 g/l |
Storage | Store at +5 to +30 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...