Thymoquinone ≥98%

Supplier: Cayman Chemical

15039-500 15039-1 15039-5
CAYM15039-500EA 52.5 EUR
CAYM15039-500 CAYM15039-1 CAYM15039-5
Thymoquinone ≥98%
Thymoquinone

The black seeds of Nigella have long been ingested to treat a broad range of diseases, including inflammation and cancer.


Thymoquinone is a natural phytochemical isolated from the seeds of N. sativa. It has anti-inflammatory effects, blocking the synthesis of leukotrienes. Thymoquinone inhibits the growth of leukemia cells (IC50 = 1,5 μg/ml), inducing apoptosis by down-regulating the expression of Bcl-2 while up-regulating Bax expression. In addition to blocking cell proliferation and triggering apoptosis, thymoquinone (15 μM) suppresses STAT3-regulated gene expression and prevents angiogenesis.

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00001602
CAS Number: 490-91-5

Order Now

Specification Test Results

Purity ≥98%
Formal Name 2-methyl-5-(1-methylethyl)-2,5-cyclohexadiene-1,4-dione
Synonyms NSC 2228
Formulation A Crystalline Solid
λmax 253 nm
SMILES O=C(C(C(C)C)=C1)C=C(C)C1=O
InChI Code 1S/C10H12O2/c1-6(2)8-5-9(11)7(3)4-10(8)12/h4-6H,1-3H3
InChI Key KEQHJBNSCLWCAE-UHFFFAOYSA-N
Storage −20 °C
Shipping Room temperature in continental US; may vary elsewhere
Stability 2 years from the QC date provided on the Certificate of Analysis, when stored properly


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR