SIN-1 chloride ≥99%
Supplier: Cayman Chemical
82220-10
82220-100
82220-50
82220-500
CAYM82220-10EA
52.5
EUR
CAYM82220-10
CAYM82220-100
CAYM82220-50
CAYM82220-500
SIN-1 chloride ≥99%
SIN-1 chloride
As the active metabolite of molsidomine, SIN-1 (chloride) is a potent vasorelaxant and inhibitor of platelet aggregation. It produces both NO and superoxide and can therefore be used to generate peroxynitrite under physiological conditions.
Formula:
C₆H₁₁ClN₄O₂ |
CAS Number:
16142-27-1 |
Specification Test Results
Purity | ≥99% |
Formal Name | 5-amino-3-(4-morpholinyl)-1,2,3-oxadiazolium chloride |
Synonyms | Linsidomine, 3-Morpholino-sydnonimine |
Formulation | A Crystalline Solid |
λmax | 253, 296 nm |
SMILES | NC1=C[N+](N2CCOCC2)=NO1.[Cl-] |
InChI Code | 1S/C6H11N4O2.ClH/c7-6-5-10(8-12-6)9-1-3-11-4-2-9;/h5H,1-4,7H2;1H/q+1;/p-1 |
InChI Key | ZRFWHHCXSSACAW-UHFFFAOYSA-M |
Storage | −20 °C |
Stability | 2 years from the QC date provided on the Certificate of Analysis, when stored properly |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...