Anti-SPN Mouse Monoclonal Antibody (Biotin) [clone: DF-T1]
Supplier: Biotium
BNCB0027-500 BNCB0027-100
BNCB0027-500EA 560 EUR
BNCB0027-500 BNCB0027-100
Anti-SPN Mouse Monoclonal Antibody (Biotin) [clone: DF-T1]
Antibodies
This antibody recognizes a cell surface glycoprotein of 95/115/135 kDa (depending upon the extent of glycosylation), identified as CD43 [Workshop IV]. Epitope of MAb Bra7G is clearly different from that of MAb DF-T1, called b as opposed to a for DF-T1. 70-90% of T-cell lymphomas and from 22-37% of B-cell lymphomas express CD43. No reactivity has been observed with reactive B-cells. So a B-lineage population that co-expresses CD43 is highly likely to be a malignant lymphoma, especially a low-grade lymphoma, rather than a reactive B-cell population. When CD43 antibody is used in combinationMore Product Information