Anti-FN1 Mouse Monoclonal Antibody (Biotin) [clone: 2755-8]

Supplier: Biotium
BNCB0091-500 BNCB0091-100
BNCB0091-500EA 560 EUR
BNCB0091-500 BNCB0091-100
Anti-FN1 Mouse Monoclonal Antibody (Biotin) [clone: 2755-8]
Antibodies
Fibronectin is an extracellular matrix glycoprotein present on most cell surfaces, in extracellular fluids and in plasma. A high molecular weight heterodimeric protein, it was originally discovered as a protein missing from the surfaces of virus-transformed cells, and it has been shown to be involved in various functions including cell adhesion, cell motility and wound healing. Alternative splicing and glycosylation give rise to several different forms of Fibronectin, some of which exhibit restricted tissue distribution or association with malignancies. It has been shown that Myofibroblasts phenotype formation correlates with the occurrence of glycosylated Fibronectin and Fibronectin splice variants in Dupuytren's disease.

Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: 2755-8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR