Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF488A) [clone: NM106]

Supplier: Biotium
BNC880106-500 BNC880106-100
BNC880106-500EA 560 EUR
BNC880106-500 BNC880106-100
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF488A) [clone: NM106]
Antibodies
This MAb is an excellent marker for all nuclei in cells in tissues. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. This MAb recognizes an antigen associated with the nuclei in all cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in subcellular fractions. It produces a speckled pattern in normal and malignant cells and may be used to stain the nuclei of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.

Type: Primary
Antigen: Human Nuclear Antigen
Clonality: Monoclonal
Clone: NM106
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR