Anti-Hepatocyte Specific Antigen Mouse Monoclonal Antibody (CF647) [clone: HSA133]

Supplier: Biotium
BNC470133-500 BNC470133-100
BNC470133-500EA 560 EUR
BNC470133-500 BNC470133-100
Anti-Hepatocyte Specific Antigen Mouse Monoclonal Antibody (CF647) [clone: HSA133]
MAb HSA133 stains human liver canaliculi and a subset of hepatocellular carcinomas. In frozen sections, it stains liver canaliculi strongly and may be used as a marker of this hepatic substructure. Cell preparations of hepatocellular carcinoma biopsies and cell lines are found to bind this MAb on the cell surface. HSA133 strongly stains liver canaliculi and hepatic carcinoma cells using frozen sections or paraformaldehyde fixed cell preparations.

Type: Primary
Antigen: Hepatocyte Specific Antigen
Clonality: Monoclonal
Clone: HSA133
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR