Anti-CD66 molecule Mouse Monoclonal Antibody (Purified) [clone: C66/195]
Supplier: Biotium
BNUB0195-500 BNUB0195-100
BNUB0195-500EA 511 EUR
BNUB0195-500 BNUB0195-100
Anti-CD66 molecule Mouse Monoclonal Antibody (Purified) [clone: C66/195]
Antibodies
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb reacts with nonspecific cross-reacting antigen (NCA) and shows a cross-reaction with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detectingMore Product Information