Anti-HLA-DPB1 Mouse Monoclonal Antibody (CF405S) [clone: BRA-FB6]

Supplier: Biotium
BNC040199-500 BNC040199-100
BNC040199-500EA 560 EUR
BNC040199-500 BNC040199-100
Anti-HLA-DPB1 Mouse Monoclonal Antibody (CF405S) [clone: BRA-FB6]
Antibodies
Recognizes a non-polymorphic determinant of DP-MHC class II. MHC class II antigens are transmembrane glycoproteins of non-covalently linked α (33-35 kDa) and β (27-30 kDa) chains. It reportedly reacts with B- & non-T, non-B cell lines but not with T- and myeloid cell lines and leukemias. Differential expression of MHC class II antigens on fetal and adult lymphocytes, malignant B cells appears to reflect the stage of cell differentiation which may be useful in the study of lymphoproliferative disorders.

Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: BRA-FB6
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR