Anti-Phosphotyrosine Mouse Monoclonal Antibody (Purified) [clone: PY20]

Supplier: Biotium
BNUB0232-500 BNUB0232-100
BNUB0232-500EA 511 EUR
BNUB0232-500 BNUB0232-100
Anti-Phosphotyrosine Mouse Monoclonal Antibody (Purified) [clone: PY20]
Antibodies
Protein phosphorylation is a fundamental event in the regulation of a large number of intracellular processes. Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be auto-phosphorylated kinases or certain cellular protein substrates. Tyrosine-phosphorylated proteins are involved in signal transduction and in the regulation of cell proliferation. Antibody to phosphotyrosine provides an excellent tool for the detection, characterization, and purification of phosphotyrosine containing proteins. This MAb shows no cross-reaction with other phosphoamino acids and is superb for multiple applications including staining of formalin/paraffin tissues.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: PY20
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR