Anti-TAGLN Mouse Monoclonal Antibody (CF405S) [clone: TAGLN/247]

Supplier: Biotium
BNC040247-500 BNC040247-100
BNC040247-500EA 560 EUR
BNC040247-500 BNC040247-100
Anti-TAGLN Mouse Monoclonal Antibody (CF405S) [clone: TAGLN/247]
Antibodies
This MAb recognizes a 22 kDa protein, identified as Transgelin, also designated SM22-alpha. It may cross-react with SM22-beta. Transgelin is expressed abundantly in smooth muscle cells. The human transgelin gene encodes a 201 amino acid protein that contains nuclear factor-binding motifs known to regulate transcription in smooth muscle. During embryogenesis, transgelin is expressed in smooth, cardiac and skeletal muscle, but is restricted during late fetal development and adulthood to all vascular and visceral smooth muscle cells and low levels of expression in heart. Transgelin is down regulated in several transformed cell lines, indicating that a reduction of transgelin expression may be an early indicator of the onset of transformation. Transgelin also binds Actin, causing Actin fibers to gel within minutes of binding. Binding of transgelin to Actin occurs at a ratio of 1:6 Actin monomers.

Type: Primary
Antigen: TAGLN
Clonality: Monoclonal
Clone: TAGLN/247
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Human, Mouse, Pig, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR