Anti-KRT77 Mouse Monoclonal Antibody (CF405S) [clone: AE-1]

Supplier: Biotium
BNC040253-500 BNC040253-100
BNC040253-500EA 560 EUR
BNC040253-500 BNC040253-100
Anti-KRT77 Mouse Monoclonal Antibody (CF405S) [clone: AE-1]
Antibodies
This MAb recognizes the 56.5 kDa (CK10); 50 kDa (CK14); 50 kDa (CK15); 48 kDa (CK16); 40 kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40 kDa. MAb AE3 recognizes the 65-67, 64, 59, 58, 56, and 52 kDa keratins of basic subfamily. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. AE1/AE3 is a broad spectrum anti pan-keratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer.

Type: Primary
Antigen: KRT77
Clonality: Monoclonal
Clone: AE-1
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Cow, Dog, Human, Monkey, Mouse, Rabbit, Rat, Turtle
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR