Anti-CD8A Mouse Monoclonal Antibody (CF594) [clone: RIV11]

Supplier: Biotium
BNC940333-500 BNC940333-100
BNC940333-500EA 560 EUR
BNC940333-500 BNC940333-100
Anti-CD8A Mouse Monoclonal Antibody (CF594) [clone: RIV11]
Antibodies
Recognizes a protein of 32 kDa, identified as CD8a (also known as CD8 chain, T cell co-receptor, Leu2, and T8). CD8 molecule consists of two chains, termed and chain, which are expressed as a disulphide-linked heterodimer or as an homodimer. CD8 is expressed on T cell subset (cytotoxic/suppressor T cells), thymocytes and NK cells. The majority of CD8 T-cells expresses CD8 as heterodimer. Some subpopulation of CD8 T cells as well as NK cells may express homodimer. CD8 functions as a co-receptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 chain (but not chain). The cytoplasmic domain of CD8 associates with p56lck tyrosine kinase.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: RIV11
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR