Anti-Histiocytoma Marker Mouse Monoclonal Antibody (Biotin) [clone: D11]

Supplier: Biotium
BNCB0348-500 BNCB0348-100
BNCB0348-500EA 560 EUR
BNCB0348-500 BNCB0348-100
Anti-Histiocytoma Marker Mouse Monoclonal Antibody (Biotin) [clone: D11]
Antibodies
In Western blotting, this antibody detects an antigen of 125 kDa in human liver and 135 kDa in tumors of histiocytic origin. Comparative study of this MAb and a standard CD68 MAb showed that their antigens are different. Its antigen in all macrophage types studied is located on the plasma membrane and within cytoplasmic structures including lysosomes. This MAb shows a restricted reactivity to cells of the monocyte/macrophage system. It specifically reacts with blood monocytes and stains resident macrophages in a wide variety of human tissues. This MAb does not stain antigen-presenting cells,More Product Information
Order Now

SPECIFICATIONS

Antigen name Histiocytoma Marker
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Biotin
Clone Clone D11
Reactivity Human
Host Mouse
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR